Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask
The Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask is a plastic-coated single neck flask useful as a rotary evaporator condensate collection flask or as a replacement for European designation KS 20/35.
$72.90 - $218.50
Specifications
Bottom Shape | Round |
---|---|
Material | Borosilicate Glass 3.3 |
Sterility | Non-sterile |
Surface Treatment | Plastic Coated |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
01-183-000
|
DWK Life Sciences
KC25330100 |
100 mL |
Case for $72.90
|
|
01-183-001
|
DWK Life Sciences
KC25330250 |
250 mL |
Case for $93.70
|
|
01-183-002
|
DWK Life Sciences
KC25330500 |
500 mL |
Case for $111.80
|
|
01-183-003
|
DWK Life Sciences
KC253302000 |
2000 mL |
Case for $218.50
|
|
12-141-339
|
DWK Life Sciences
KC253301000 |
1000 mL |
Case for $199.50
|
|
Description
- Spherical socket joint
- Joint Size: 35/20 mm
- Autoclavable KimCote safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Round | |
Borosilicate Glass 3.3 | |
Non-sterile | |
Plastic Coated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title