Learn More
DWK Life Sciences Kimble™ KIMAX™ Petri Dishes Set Available on GSA/VA Contract for Federal Government customers only.
Designed from Federal Specification NNN-D-1478.
$747.24
Specifications
Edges Reinforced Beaded
Material Borosilicate Glass
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Price Quantity & Availability  
08-746B Available on GSA/VA Contract for Federal Government customers only.
View Documents
DWK Life Sciences
2306010010
100 mm 10 mm
Case of 72 for $747.24
Add to cart
Description
Description
  • Reinforced beaded edges resist mechanical breakage and to help in centering the bottom inside the cover
  • Remain clear after repeated sterilization
  • Covers with fused-on red enamel marking spot and inscription
  • Bottoms with white enamel inscription and vertical arrow on side to locate starting point for serial dilutions
  • Bottoms have a vertical arrow on the side to precisely locate serial dilution starting points
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Reinforced Beaded
Borosilicate Glass