Learn More
DWK Life Sciences Kimble™ KIMAX™ Petri Dishes Set
Designed from Federal Specification NNN-D-1478.
Manufacturer:  DWK Life Sciences 2306010010
 View more versions of this product
Catalog No. 08-746B
Add to cart
Description
Description
  • Reinforced beaded edges resist mechanical breakage and to help in centering the bottom inside the cover
  • Remain clear after repeated sterilization
  • Covers with fused-on red enamel marking spot and inscription
  • Bottoms with white enamel inscription and vertical arrow on side to locate starting point for serial dilutions
  • Bottoms have a vertical arrow on the side to precisely locate serial dilution starting points
Specifications
Specifications
100 mm
Borosilicate Glass
10mm
Reinforced Beaded