Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Borosilicate Glass Tubes with Plain End
Without marking spot
Supplier: DWK Life Sciences 4506025150
Description
Without marking spot
- Culture tubes made from KG-33 glass feature fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance
- Designed from ASTM™ specification E982, Type IV requirements
Specifications
ASTM E982, Type IV | |
55 mL | |
Borosilicate Glass | |
25 mm |
Plain | |
150 mm | |
None |