Learn More
DWK Life Sciences Kimble™ Kimcote™ Heavy Wall Recovery Flask
Plastic-coated single neck flask with a standard taper outer joint designed for easy recovery of reaction products
Manufacturer:  DWK Life Sciences KC25165200
 View more versions of this product
Catalog No. 12-141-501
Add to cart
Description
Description
  • Autoclavable KimCote™ safety coating reduces hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Even wall thickness and minimum runout make flasks ideal for use with rotary evaporators
  • Heavy wall square joint bead
  • Total contents accessible with a spatula or policeman
  • Lower portion fits standard heating mantles
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
144 mm
24/40 mm
76 mm
200 mL
Heavy wall recovery flask
Autoclavable