Learn More
DWK Life Sciences Kimble™ Kontes™ Distilling Three Neck Round Bottom Heavy Wall Flask
Side necks are angled
Manufacturer:  DWK Life Sciences 6060200224
 View more versions of this product
Catalog No. K606020-0224
Add to cart
Description
Description
  • Heavy wall reduces breakage.
Specifications
Specifications
2.36 in.
Borosilicate Glass
3
Round
100 mL
Angled
24/40 Center, 24/40 side