Learn More
DWK Life Sciences Kimble™ Kontes™ Erlenmeyer Flasks
Used to measure, mix, and store liquids
Supplier: DWK Life Sciences 6170000124
Description
Used to measure, mix, and store liquids
- Single neck flask
- Has standard taperouter joint
Specifications
50 mL | |
3.5 in. | |
24/40 |
Borosilicate Glass | |
9 cm | |
Narrow |