Learn More
DWK Life Sciences Kimble™ Kontes™ Hose Inlet Adapters
For use as a vent or for the introduction of pressure/vacuum
$58.60 - $77.50
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
View More Specs
Products 7
Catalog Number Mfr. No. Standard Taper Joint No. Fits Tubing ID (English) Height (English) Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint No. Fits Tubing ID (English) Height (English) Product Type Price Quantity & Availability  
K276100-1922
View Documents Promotion Details
DWK Life Sciences
2761001922
19/22 0.25 in. 2.55 in. Inner Joint Hose Inlet Adapter
Case for $62.80
Add to Cart
K276110-1420
View Documents Promotion Details
DWK Life Sciences
2761101420
14/20 0.25 in. 2.36 in. Outer Joint Hose Inlet Adapter
Case for $60.70
Add to Cart
K182910-2440
View Documents Promotion Details
DWK Life Sciences
1829102440
24/40 0.37 in. 3.34 in. Outer Joint Hose Inlet Adapter
Case for $77.50
Add to Cart
K276000-0000
View Documents Promotion Details
DWK Life Sciences
2760000000
14/20 0.25 in. 1.77 in. 90° Hose Inlet Adapter
Case for $58.60
Add to Cart
K276010-0000
View Documents Promotion Details
DWK Life Sciences
2760100000
19/22 0.25 in. 2.16 in. 90° Hose Inlet Adapter Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. N/A
K276100-1420
View Documents Promotion Details
DWK Life Sciences
2761001420
14/20 0.25 in. 2.36 in. Inner Joint Hose Inlet Adapter Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
K183000-2942
View Documents Promotion Details
DWK Life Sciences
1830002942
29/42 0.37 in. 3.34 in. 90° Hose Inlet Adapter Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
Description
Description
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Borosilicate Glass