Learn More
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
Supplier:  DWK Life Sciences 4225300025
 View more versions of this product
Catalog No. K422530-0025
Add to cart
Description
Description
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
  • Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
  • Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
  • Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
  • Adjust spray pattern by varying the position of thumb on vent hole
  • Autoclavable (after cap, loosening ring, and O-ring are removed)
Specifications
Specifications
Autoclavable
25 mL
Glass
TLC Reagent Sprayer
Standard TLC Reagents and Most Corrosive Liquids
19/22
14.7 cm
4 cm
0.84 oz.
Screw thread ground joint, connecting cap, loosening ring
1 Case
To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography
3 to 5 psi
5.78 in.
1.57 in.