Learn More
DWK Life Sciences Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
$138.00 - $469.00
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
K953760-1002
|
DWK Life Sciences
9537601002 |
1000 mL |
Case for $265.00
|
|
|||||
K953760-4002
|
DWK Life Sciences
9537604002 |
4 L |
Case for $469.00
|
|
|||||
K953760-2002
|
DWK Life Sciences
9537602002 |
2000 mL |
Case for $384.00
|
|
|||||
K953760-0122
|
DWK Life Sciences
9537600122 |
125 mL |
Case for $138.00
|
|
|||||
Description
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- KimCote™ plastic safety coating
- ≠2 side arm for connection to 0..25 in. (6mm) I.D. flexible tubing
- 125mL flask: With No. 5 stopper joint
- 1, 2 and 4L flasks: With No. 8 stopper joint
Specifications
Borosilicate Glass |