Learn More
DWK Life Sciences Kimble™ Low-Form Color-Comparison Tubes
SDP
Shadowless bottom construction allows for viewing with undistorted light
$319.00 - $463.00
Specifications
Material Borosilicate Glass
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Graduations Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Graduations Price Quantity & Availability  
10-310-188A
SDP
View Documents
DWK Life Sciences
4531050100
100 mL 50 and 100 mL
Case of 6 for $420.00
Add to Cart
10-310-190
SDP
View Documents
DWK Life Sciences
45310A100
100 mL 100 mL
Case of 6 for $463.00
Add to Cart
10-310-191
SDP
View Documents
DWK Life Sciences
45310A50
50 mL 50 mL
Case of 6 for $319.00
Edge
Add to Cart
Description
Description
Shadowless bottom construction allows for viewing with undistorted light
  • Rings and legend are durable white ceramic enamel
  • Rings coincide with specific volumes of liquid
  • Matched or unmatched sets of 50 and 100mL
  • Glass, nonsterile, autoclavable
Order Info
Graduated ring scale of any of the six tubes will not vary by more than 3mm from that of any other tube within the set.
Specifications
Specifications
Borosilicate Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.