Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Pulp/Vinyl Liners
Click to view available options
Neck Finish:
20-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Description
- Polyvinyl film is adhered to 0.035-in. pulpboard
- Color: black
- Low cost cap for general purpose usage
- Not recommended for hydrocarbons or bleaches
Specifications
Closure Liner | Pulp/Vinyl |
For Use With (Equipment) | Bottle |
Product Type | Phenolic Cap |
Color | Black |
Material | Phenolic Resin |
No. per Case | 144 Cs. |
Neck Finish | 38-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction