Learn More
Kimble™ Phenolic Caps with Tinfoil Liners
Manufacturer:  Kimble™ 75203G43400
 View more versions of this product
Catalog No. 13-757-147
  Check Availability
Add to cart
Alert:
Not recommended for acids and alkalis.
Specifications
Description
  • Economical general purpose phenolic screw cap with a tinfoil liner
  • Color: black
  • Best suited for environmental sampling applications
  • Foil is laminated to paper and bonded to a 0.035-in. pulpboard
Description & Specifications
Specifications
Tin Foil
Phenolic
Phenolic Caps
43-400
Black
144 Cs.
Bottle