Learn More
DWK Life Sciences Kimble™ Precision Stirrer Assembly
SDP
Recommended for reaction systems requiring a vacuum
Supplier:  DWK Life Sciences 7820300124
Catalog No. 12-141-486
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • PTFE blade
  • Complete unit includes a polyacetal stuffing box, glass stirrer bearing
  • Packing consists of a PTFE and graphite mixture
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
500 to 5000 mL
PTFE blade, Polyacetal stuffing box, glass stirrer bearing, OD glass shaft, Graphite mixture
24/40 mm
19 mm
445 mm
Blade 75 mm
Product Suggestions
Product Suggestions