Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Serum KG-35 Borosilicate Glass Aluminum Seal without Closure
Click to view available options
Capacity (Metric):
5 mL
100 mL
125 mL
200 mL
Description
Neutraglas™
- No closure
- GPI Neck Finish: 20A
Specifications
Closure Material | Aluminum |
Closure Size | 20A |
Diameter (Metric) | 52 mm |
Length (English) | 3.7 in. |
Product Type | Serum Bottle |
Capacity | 3.38 oz., 100mL |
Capacity (Metric) | 100 mL |
Capacity (English) | 3.38 oz. |
Diameter (English) | 2.04 in. |
Material | Borosilicate Glass |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction