Learn More
DWK Life Sciences Kimble™ Stainless Steel Standard Taper Joint Clamp
The Kimble™ Stainless Steel Standard Taper Joint Clamp is used to secure standard ground joint connections in glass assemblies and is characterized by its easy handling and excellent resilience.
Supplier: DWK Life Sciences 6754002440
This item is not returnable.
View return policy
Description
- Available in either metal or polyoxymethylene (POM)
- Dimensionally stable with particularly good spring properties
- Easy, safe and quick to fit
- Superior chemical and temperature resistance
- Can be heated up to 500°C
- Plastic clips are marked with their joint size and are color coded to allow easy identification of the size of clip required
Specifications
Standard Taper Joint Clamp | |
Stainless Steel | |
24/40, 24/25 |