Learn More
DWK Life Sciences Kimble™ Sterile Disposable Borosilicate Glass Milk Bacteriological Pipets
Plugged and sterile, with a uniform pulled tip
Manufacturer:  DWK Life Sciences 7210622
 View more versions of this product
Catalog No. 03-340-71G
  Check Availability
Add to cart
Description
Description
  • 1.1mL size also used for milk phosphatase test
  • Packages are color coded for easy identification by size
Order Info
Supplied in individual paper/plastic packages, which can be opened by either the snap-through or peel-apart method
Compliance
Meet APHA requirements for making milk dilutions
Specifications
Specifications
APHA, USP type I, ASTM E438, type I, Class A
Yes
Uniform pulled tip
Plugged Bacteriological Pipet
Borosilicate Glass
Yes
1, 2, 2.1, 2.2mL
±0.04mL
2.2mL
Sterile