Learn More
DWK Life Sciences Kimble™ ValueWare™ Culture Tubes with Rubber Lined Phenolic Caps
SDPSCI_ED
Mineral-filled cap to withstand repeated autoclaving.
Supplier:  DWK Life Sciences 5506616150
 View more versions of this product
Catalog No. S00206
Add to Cart
Description
Description
  • KIMAX™ KG-33 borosilicate glass
  • Uniform wall thickness for maximum heat transfer and chemical resistance
  • Screw Thread finish complies with Glass Packaging Institute (GPI) requirements
  • Cap not atttached
  • Reusable
Compliance
Designed from ASTM™ Specification E982, Type VI, Class A requirements
Specifications
Specifications
ASTM E982, Type VI, Class A
20 mL
Borosilicate Glass
Rubber
16 mm
Phenolic
150 mm
No
Autoclavable
Mineral-filled Screw Cap
Product Suggestions
Product Suggestions