Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ ValueWare™ Narrow-Mouth Erlenmeyer Flasks
Economical and versatile; ideal for general laboratory usage.
Supplier: DWK Life Sciences 5650025
Description
- Kimax™ KG-33 glass Reinforced tops tooled with a rounded finish — contain more glass for maximum mechanical strength
- Thick-walled body has long tapered outside contour to minimize chipping when struck or rubbed together
- Durable white ceramic enamel scales indicate approximate volumes at various levels — useful for measuring and mixing solutions where a high degree of accuracy is not necessary
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E1404, Type I, Class 1 specification requirements.
Specifications
Rubber | |
1.53 in. | |
Borosilicate Glass | |
5 mL | |
6.5 cm | |
3.9 cm |
White Ceramic Enamel scale graduations; Top style: Reinforced and tooled with rounded finish beaded | |
25 mL | |
10 to 25 mL | |
2.55 in. | |
Narrow | |
0 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction