Learn More
DWK Life Sciences Kimble™ ValueWare™ Petri Dishes with Covers
SCI_ED
Color-coded markings for quick and easy identification.
Specifications
Color Clear
Material Glass
Shape Round
Edges Reinforced Beaded
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Price Quantity & Availability  
S00055
SCI_ED
View Documents
DWK Life Sciences
5306010020/EMD
100 mm 20 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Kimax™ KG-33 glass remains clear after repeated wet or dry sterilization cycles
  • Reinforced beaded edges resist mechanical breakage, help in centering bottom inside cover
  • Covers have bright red ceramic enamel marking spot and COVER inscription
  • Bottoms have white ceramic enamel BOTTOM inscription and vertical arrow to precisely locate serial dilution starting points
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Dish and covers designed to Federal Specification NNN-D-1478 requirements.
Specifications
Specifications
Clear
Glass
Round
Reinforced Beaded