Learn More
Kimberly-Clark™ Kimtech™ G3 NxT Nitrile Cleanroom Gloves
Latex and silicone-free non-sterile cleanroom gloves for delicate applications. The slick finishing, with textured fingertips and static dissipative material, ensures dextrous contamination control.
Specifications
Length (Metric) 30.5 cm
Color White
Hand Ambidextrous
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19059327A
View Documents
Kimberly-Clark™
62090
X-Small N/A
19-059-327B
View Documents
Kimberly-Clark™
62091
Small N/A
19-059-327C
View Documents
Kimberly-Clark™
62092
Medium N/A
19-059-327D
View Documents
Kimberly-Clark™
62093
Large N/A
19-059-327F
View Documents
Kimberly-Clark™
62094
Large+ N/A
19-059-327E
View Documents
Kimberly-Clark™
62095
X-Large N/A
Description
Description
  • The synthetic nitrile polymer material is designed for fit and reliability, with textured fingertips for improved grip and excellent water tightness that results in a low risk of pinholes.
  • The gloves are ambidextrous and incorporate a beaded cuff for added strength and ease of donning
  • Our non-sterile nitrile cleanroom gloves are latex, silicone, powder-free and manufactured without vulcanisation accelerators
  • The absence of natural rubber latex and vulcanizing agents reduces the risk of TYPE I and TYPE IV glove-associated reactions, protecting the wearer as well as the application.
  • Kimtech™ G3 NxT Nitrile Cleanroom Gloves keep hands comfortable and protected while ensuring that research applications can be carried out contamination-free.
  • Kimberly-Clark Professional personal protective equipment is only intended for occupational workplace use and must be used in compliance with all applicable health and safety laws and standards in addition to all user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
30.5 cm
White
Ambidextrous