Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Kimtech™ Kimguard™ KC100 Sterilization Wrap

Kimtech™ Kimguard™ Sterilization Wrap is designed for sterile processing environments, such as in pharmaceuticals and medical devices.

Manufacturer:  Kimberly-Clark Professional™ 10745

 View more versions of this product

Catalog No. 19-065-700


Add to cart

Description

Description

  • The first of six strengths of Kimtech™ sterilization wrap
  • Made of low-lint SMS fabric
  • Compatible with both pre-vacuum steam sterilant penetration and EO sterilant penetration and residuals
  • Tear-resistant (Tensile Strength 12.6 lbs. ASTM D5034)
  • Flame-resistant (Class 1 flammability rating)
  • Bacterial filtration efficiency (BFE) of 96.2%
  • Maintenance of package integrity (MPI) to meet FDA requirements and instructions for use that meet FDA guidelines
  • Recyclable via The RightCycle Program by Kimberly-Clark Professional
Specifications

Specifications

Safety
Blue
45 in.
KC100
SMS
45 in.
Sterile
Sterile processing environments
114 cm
Sterilization Wrap
1000 sheets/case
114 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit