Learn More
Kimberly-Clark Professional™ Kimtech™ Kimguard™ KC600 Sterilization Wrap
Kimtech™ Kimguard™ Sterilization Wrap is designed for sterile processing environments, such as in pharmaceuticals and medical devices.
Manufacturer:  Kimberly-Clark Professional™ 68648
Catalog No. 19-169-986
Add to cart
Description
Description
  • Steam and EO compatible Industry standard for sterile processing
  • Tear-, flame-, lint- and abrasion-resistant
  • Powerguard™ technology for enhanced microbial barrier protection
  • Pre-vacuum steam sterilant penetration Ethylene Oxide (EO) sterilant penetration and residuals
  • Maintenance of package integrity (MPI) to meet FDA requirements
  • Bacterial Filtration Efficiency (BFE) from 98.9% to 99.9%
  • Instructions for Use that meet FDA guidelines Class 1 flammability rating
  • Recyclable via The RightCycle Program by Kimberly-Clark Professional
Specifications
Specifications
Sterile
48 x 48 in.
48 in.
KC600
48 in.
121 cm
Blue
Sterile processing environments
121 cm
Sterilization Wrap
Low-lint SMS