Learn More
Kimberly-Clark™ Professional Kimtech Pure™ G3 Nitrile Gloves
Offer advanced levels of process protection and comfort in ISO 3 or higher cleanrooms.
Manufacturer:  Kimberly-Clark™ Professional HC61010LC
Catalog No. 19-048-653ALC
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Clean processed for low particle and extractable levels
  • Ambidextrous with beaded cuff for added strength
  • Textured fingertips for improved grip
  • Silicone-free formulation
  • Double-lined bags
Specifications
Specifications
Safety
Textured fingertips
Ambidextrous
Nitrile gloves
White
Silicone free formulation
Beaded/Rolled
X-Small
G3