Learn More
Kimberly-Clark Professional™ Kimtech™ Pure G5 Latex 10 in. Cleanroom Gloves
G5 non-sterile latex gloves measure 10 in. in length and feature textured palms and fingertips for a secure grip. Recommended for Class 100/ISO 5 or higher critical environments.
Specifications
Certifications/Compliance ASTM D3578
Cleanroom Class Class 100/ISO 5
AQL 1.5
Thickness (English) 8 mil
Thickness (Metric) 0.20 mm
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
11-390-4A Greener Choice
View Documents
Kimberly-Clark Professional™
56805
Small
Pack of 100 for $35.54
Case of 10 PK for $355.41
N/A
Due to product restrictions, please to purchase or view availability for this product.
11-390-4B Greener Choice
View Documents
Kimberly-Clark Professional™
56806
Medium
Pack of 100 for $35.54
Case of 10 PK for $355.41
N/A
Due to product restrictions, please to purchase or view availability for this product.
11-390-4C Greener Choice
View Documents
Kimberly-Clark Professional™
56807
Large
Pack of 100 for $35.54
Case of 10 PK for $355.41
N/A
Due to product restrictions, please to purchase or view availability for this product.
11-390-4D Greener Choice
View Documents
Kimberly-Clark Professional™
56808
X-Large
Pack of 100 for $35.54
Case of 10 PK for $355.41
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Low in natural rubber proteins
  • Textured palm and fingers
  • Beaded cuff adds strength and makes donning easier
  • Ambidextrous
  • Thickness: 8.3 mil
  • Length: 10 in. (25 cm)
Specifications
Specifications
ASTM D3578
1.5
0.20 mm
25.5 cm
Class 100/ISO 5
8 mil
10 in.
No Texture