Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565220
Description
Immunogen sequence: IEEQVRRGLEG KEQEVPTFTE LSRENDEEKV TFNLSKGACS SSGATSSKSS TLGPSALKTI GSSASVKRKE SS Highest antigen sequence identity to the following orthologs - mouse 82%, rat 85%.
Kin is a nuclear protein that forms intranuclear foci during proliferation and is redistributed in the nucleoplasm during the cell cycle. Short-wave ultraviolet light provokes the relocalization of the protein, suggesting its participation in the cellular response to DNA damage. Originally selected based on protein-binding with RecA antibodies, the mouse protein presents a limited similarity with a functional domain of the bacterial RecA protein, a characteristic shared by the human ortholog.
Specifications
KIN | |
Polyclonal | |
Unconjugated | |
Kin | |
antigenic determinant of rec-A protein; antigenic determinant of rec-A protein homolog; binding to curved DNA; Btcd; DNA/RNA-binding protein KIN17; HsKin17 protein; KIN; KIN, antigenic determinant of recA protein; KIN, antigenic determinant of recA protein homolog; KIN17; Kin17 DNA and RNA binding protein; Rts2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22944 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60870 | |
Kin | |
Recombinant Human KIN. Recombinant protein control fragment (Product #RP-104693). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction