Learn More
Alfa Aesar™ Kinase buffer V (5X)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J62045AK
 View more versions of this product
Catalog No. AAJ62045AK
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • Formulation: 100mM MOPS, 50mM magnesium chloride, and 50mM manganese chloride, pH 7.4
Specifications
Specifications
Kinase buffer V (5X)
Liquid
100mM MOPS, 50mM magnesium chloride, and 50mM manganese chloride, pH 7.4
250mL