Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kindlin Mouse anti-Human, Clone: 4A5.14, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Kindlin
Clone 4A5.14
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug of protein 1ml of lysate, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122865
View Documents
Novus Biologicals
NBP30769610UG
100 μg
Each for $352.89
Add to cart
 
NB122866
View Documents
Novus Biologicals
NBP30769620UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Kindlin Monoclonal antibody specifically detects Kindlin in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

Kindlin
0.2 mg/ml
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
55612
IgG2a Lambda
Primary
Store at 4°C.
4A5.14
Western Blot 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug of protein 1ml of lysate, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human
C20orf42fermitin family homolog 1, chromosome 20 open reading frame 42, fermitin family homolog 1 (Drosophila), fermitin family member 1, FLJ20116, KIND1DTGCU2, kindlerin, kindlin 1, Kindlin syndrome protein, Kindlin-1, UNC112 related protein 1, UNC112A, Unc-112-related protein 1, URP1FLJ23423
Recombinant protein containing Kindlin (residues 216-677)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit