Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
kinectin Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232262AF750
Description
kinectin Polyclonal antibody specifically detects kinectin in Human samples. It is validated for Western BlotSpecifications
kinectin | |
Polyclonal | |
Western Blot | |
kinectin, kinectin 1 (kinesin receptor), MGC133337 | |
The immunogen this antibody was made to, maps to a region between residue 1307 to 1357 of human Kinectin using the numbering given in entry NP_001072989.1 (GeneID 3895). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
3895 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction