Learn More
Kinematica HMC Europe Vertical Autoclave Air Filters
Exhaust air filter. Safety for individuals and the environment. Option for safe, emission-free sterilization of highly infectious germs and genetically modified materials. Not for applications requiring FDA approval.
$614.00 - $9110.00
Specifications
For Use With (Application) Sterilization of exhaust air
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity & Availability  
08-451-409
View Documents
Kinematica HMC Europe
51011
HMC-HG autoclave Exhaust Air Filter
Each for $9,110.00
Only null left
Add to Cart
08-451-435
View Documents
Kinematica HMC Europe
50019A
HMC-HV autoclave Replacement Air Filter
Each for $614.00
Only null left
Add to Cart
08-451-434
View Documents
Kinematica HMC Europe
50004
HMC-HV autoclave Exhaust Air Filter
Each for $8,640.00
Only null left
Add to Cart
Description
Description
During the venting phase, the steam displaces unsterilized residual air from the chamber and passes it through a filter system. The surrounding air is protected against contamination as the cycle is running. Naturally, in-line sterilization of the exhaust air filter is performed. Due to the design, condensate sterilization is a matter of course. Any condensate which precipitates flows back into the chamber and gets sterilized. Discharge can be done safely.
Specifications
Specifications
Sterilization of exhaust air
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.