KINEMATICA Polytron Dispersing Aggregates: for PT-3100 and PT-3100D Homogenizers

Manufacturer: KINEMATICA 11030144

 View all options for this product

Catalog No. 08-451-100


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Type Dispersing Aggregates
    For Use With (Equipment) PT 3100 D homogenizers
    Model PT-DA 36/2EC-B250
    Sample Volume (English) 200 to 4000mL
    Speed 30 m/s

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit