Learn More
Kinematica Polytron Generator: for PT-3100 and PT-3100D Homogenizers
Outside blades pre-crush samples that are larger than the rotors diameter. Subsequently, they can be dispersed inside the generator.
Supplier: Kinematica 11030048
Description
Applications:
- Dispersing of plant, human or animal tissues, resins and pigments
- Extractions
- Producing of suspensions and emulsions
Specifications
Dispersing Aggregates | |
2 to 250 mL | |
PT 3100 homogenizers |
13.5 m/s | |
170 mm | |
PT-DA 3012/2M |