Learn More
Kinematica Polytron™ Generators
For Benchtop rotor/stator homogenizers
Manufacturer:  Kinematica 11030074
 View more versions of this product
Catalog No. 08-451-42
Add to cart
Description
Description
Ideal when cross-contamination is a concern and cleaning is necessary.
  • One-piece Type 316L stainless-steel
  • Stronger, thicker-walled stator tube that extends generator life and prevents deformation
  • Controlled tolerances ensure a small and uniform rotor-stator gap for maximum shear force and reproducible particle yield
  • “Quick-connect” generators connect easily for quick changes Saw teeth to facilitate processing and foam-reducing features to reduce air intake
  • EasyCare™ generators disassemble for cleaning, maintenance and sterilization
Generators with knives are used for processing samples that require precutting and are larger than generator diameter.
Specifications
Specifications
10 to 20000mL
20.5cm
Ideal for dispersing of plant, human or animal tissues, resins and pigments, producing of suspensions and emulsions
8.07 in.
PT DA 3020/2
Standard, long shaft slots, 1 rotating, 1 stationary ring
Generator
Polytron™ PT 3100/6100 homogenizer
Stainless Steel