Learn More
Kinematica Polytron Generators
For Benchtop rotor/stator homogenizers
$2773.00
Catalog Number | Mfr. No. | Capacity (Metric) | Length (Metric) Shaft | For Use With (Equipment) | Length (English) Shaft | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Length (Metric) Shaft | For Use With (Equipment) | Length (English) Shaft | Price | Quantity & Availability | ||
08-451-42
|
Kinematica
11030074 |
10 to 20000mL | 20.5cm | Polytron™ PT 3100/6100 homogenizer | 8.07 in. |
Each for $2,773.00
|
|
Description
Ideal when cross-contamination is a concern and cleaning is necessary.
- One-piece Type 316L stainless-steel
- Stronger, thicker-walled stator tube that extends generator life and prevents deformation
- Controlled tolerances ensure a small and uniform rotor-stator gap for maximum shear force and reproducible particle yield
- “Quick-connect” generators connect easily for quick changes Saw teeth to facilitate processing and foam-reducing features to reduce air intake
- EasyCare generators disassemble for cleaning, maintenance and sterilization
Generators with knives are used for processing samples that require precutting and are larger than generator diameter.
Specifications
Generator | |
Stainless Steel |