KINEMATICA Polytron™ PT 1600E Benchtop Homogenizers
Fast and successful sample preparation for small-scale applications
Manufacturer: KINEMATICA 11090005
 View more versions of this product
Catalog No. 08-451-168
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Quantity Each
    Includes Drive unit model 1600 E, plate stand model PT ST 1603, vessel holder with boss head and safety positioning ring
    Product Type Homogenizer Package
    The homogenizer processes emulsions, suspensions and foams by reducing the size of solid particles, droplets and gas bubbles to a few microns or less. Small and large amounts of product are dispersed quickly and efficiently.
    • Soft-start with 300w powerful motor
    • Rotor speed range from 5000 up to 30,000rpm
    • Tip speeds up to approx. 14m/s
    • Integrated electronic speed regulation/stabilization with control knob and speed scale
    • Quick coupling for dispersing aggregates and to change heads scatterers without the need for tools
    • Four different, exchangeable dispersing aggregates in EasyClean EC-design
    Note: Homogenizer does not come with aggregates
  • Description & Specifications
Related Products
Product Content Feedback