Learn More
Kinematica Polytron™ Model PT 10-35 GT / PT 3100 D Homogenizer Accessory, Generator
Powerful and versatile benchtop homogenizer. KINEMATICA Polytron™ Model PT 10-35 GT-D Homogenizer completely homogenizes samples from 0.1 to 4000mL.
Manufacturer:  Kinematica 11030112
 View more versions of this product
Catalog No. 08-451-212
Add to cart
Description
Description
  • Completely homogenizes samples from 0.1 to 4000mL
  • Powerful 1200W motor provides quiet and reproducible results
  • Homogenize small tissues to fine emulsions and suspensions
  • Get the right results with a variety of generators
Specifications
Specifications
Z Stator
PT 10-35 GT and PT 3100 D
100 to 3000mL
PT-DA 30/2ZEC-B250