Learn More
Kinematica Polytron™ PT1200E Handheld Homogenizer
Lightweight model breaks your samples down fast while offering outstanding durability
$2440.00
Specifications
For Use With (Application) Ideal for pulverizing plant and animal tissue, grain and seed, soil extraction, DNA/RNA extraction, Karl Fischer sample prep, toxicology and food science
Model PT 1200E
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
05-400-261
Accessories View Documents
Kinematica
11010026
Homogenizer; 115V Each for $2,440.00
Add to cart
Description
Description
  • Ergonomic design for repetitive motions and prolonged use
  • Homogenization of plant and animal tissues
  • 100 W motor with safety power supply, fingertip speed control to 25,000 rpm,
  • Quick-connect coupling makes changing generators easy
  • Full line of stainless steel and disposable generators
  • Accommodates samples up to 250mL
Specifications
Specifications
Ideal for pulverizing plant and animal tissue, grain and seed, soil extraction, DNA/RNA extraction, Karl Fischer sample prep, toxicology and food science
PT 1200E