Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin 5B Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500580IR
Description
Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human, Mouse, Porcine samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Kinesin 5B | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Porcine | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 755 | |
Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
Stalk domain of human kinesin (amino acids 331-906) expressed in E. coli (FKHC3). | |
0.1 mL | |
Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
3799 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction