Learn More
Thermo Scientific™ KingFisher™ Cell and Tissue DNA Kit Non-distribution product as customer accommodation.
Rapidly purify high-quality, reproducible DNA from cell and tissue samples and cultured bacteria with this unique nucleic acid purification workflow.
Manufacturer:  Thermo Scientific™ 97030496
 View more versions of this product
Catalog No. 97-030-496
Add to cart
Description
Description
Rapidly purify high-quality, reproducible DNA from cell and tissue samples, as well as cultured bacteria, with the Thermo Scientific™ KingFisher™ Cell and Tissue DNA Kit. This unique nucleic acid purification workflow provides an optimized, high-throughput method for extreme flexibility.
Features:
  • Purifies DNY from maximum 1 x 107 cells or 20mg of tissue or 1mL of cultured bacteria
  • Produces isolated DNA that is free of inhibitors and ready for use in many downstream applications
  • Includes easy-to-use protocols for safe sample-handling as well as minimal user input, maximizing walkaway time
  • Optimized to produce high yields of quality DNA
  • Suitable for wide range of sample materials from cells to tissues
  • Flexible throughput from one sample to up to 96 samples per run
  • Compatible with most common downstream analysis and applications
Recommended for:
Genomics, target identification, veterinary assays, biomarker discovery, quality control
Specifications
Specifications
Tissues, cultured cells or bacteria
384 samples
KingFisher Cell and Tissue DNA Kit; 384 samples
25 min.
Up to 20mg of tissue, 1 x 107 cells, or 1mL of overnight cultured bacteria
PCR, real-time PCR, enzymatic reactions
100µL (Duo) or 150µL (Flex)