Thermo Scientific™ KingFisher™ Pure DNA Blood Kit Available on GSA/VA Contract for Federal Government customers only.

Achieve rapid, reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate with this unique nucleic acid purification workflow.

Manufacturer: Thermo Scientific™ 98010496

 View more versions of this product

Catalog No. 22-387-063

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Sample Type Fresh or frozen whole blood treated with EDTA or citrate
    Sample Input 200µL or 1mL
    Kit Size 384 samples
    Suitable Downstream Analysis PCR, real-time PCR, enzymatic reactions
    Description KingFisher Pure DNA Blood Kit; 384 samples
    Elution Volume (Metric) 200µL sample: 100µL (Duo) or 150µL (Flex). 1mL sample: 500µL. Recommended but adjustable
    Run Time 40 min. (200µL), 45 min. (1mL)

    Achieve rapid, reproducible purification of high-quality DNA. The Thermo Scientific™ KingFisher™ Pure DNA Blood Kit, together with a KingFisher magnetic particle processor, allows rapid and reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate. Thermo Scientific KingFisher Kits complete the unique nucleic acid purification workflow providing an optimized high throughput method for extreme flexibility.

    Features

    • Produces isolated DNA that is free of inhibitors and ready for use in many downstream applications
    • Easy-to-use protocols for safe sample-handling, as well as minimal user input, maximizing walkaway time.
    • Excellent performance and consistency of results optimized to produce high yields of quality DNA
    • Large volume sample input possible up to 3mL of blood per sample
    • Flexible throughput from one sample to up to 96 samples per run
    • Minimized hands-on time increases efficiency
    • Compatible with most common downstream analysis and applications

    Recommended for:

    Genomics, target identification, veterinary assays, biomarker discovery, quality control

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit