Thermo Scientific™ KingFisher™ Pure Plasmid Kit Available on GSA/VA Contract for Federal Government customers only.
Achieve rapid and reproducible purification of high-quality plasmid DNA from cultured bacteria.
Manufacturer: Thermo Scientific™ 98080496
 View more versions of this product
Catalog No. 22-387-074
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Description KingFisher Pure Plasmid Kit, 384 samples
    Sample Type Bacterial culture
    Sample Input Up to 5mL
    Kit Size 384 samples
    Suitable Downstream Analysis PCR, real-time PCR, enzymatic reactions
    Elution Volume (Metric) 100µL
    Run Time 40 min.
    Thermo Scientific™ KingFisher ™Pure Plasmid Kit is designed for rapid automated purification of plasmid DNA from cultured bacteria using KingFisher instruments. Plasmid DNA can be purified from 1-5mL of bacterial culture. The excellent performance of KingFisher Plasmid DNA Kit produces plasmid DNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
    • Excellent yield, purity and reproducibility
    • Flexible KingFisher system
    • Walk-away solution for any throughput
    • Compatible with most common downstream analysis and application
  • Description & Specifications
Related Products
Product Content Feedback