Learn More
Thermo Scientific™ KingFisher™ Pure Plasmid Kit
Achieve rapid and reproducible purification of high-quality plasmid DNA from cultured bacteria.
Manufacturer:  Thermo Scientific™ 98080196
 View more versions of this product
Catalog No. 22-387-073
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ KingFisher ™Pure Plasmid Kit is designed for rapid automated purification of plasmid DNA from cultured bacteria using KingFisher instruments. Plasmid DNA can be purified from 1-5mL of bacterial culture. The excellent performance of KingFisher Plasmid DNA Kit produces plasmid DNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
  • Excellent yield, purity and reproducibility
  • Flexible KingFisher system
  • Walk-away solution for any throughput
  • Compatible with most common downstream analysis and application
Specifications
Specifications
Bacterial culture
96 samples
KingFisher Pure Plasmid Kit, 96 samples
40 min.
Up to 5mL
PCR, real-time PCR, enzymatic reactions
100µL