Thermo Scientific™ KingFisher™ Pure RNA Plant Kit Available on GSA/VA Contract for Federal Government customers only.
Achieve rapid and reproducible purification of high-quality RNA from various plant materials.
Manufacturer: Thermo Scientific™ 98060196
 View more versions of this product
Catalog No. 22-387-069
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Sample Type Fresh or frozen plant tissues
    Sample Input Up to 50mg of fresh plant tissue
    Kit Size 96 samples
    Suitable Downstream Analysis PCR, real-time PCR, enzymatic reactions
    Description KingFisher Pure RNA Plant Kit; 96 samples
    Elution Volume (Metric) 100µL
    Run Time 60 min.
    Thermo Scientific™ KingFisher™ Pure RNA Plant Kit is designed for rapid automated purification of total RNA from plant samples using KingFisher instruments. Total RNA can be purified from up to 50mg of fresh plant tissue. The excellent performance of KingFisher Pure RNA Plant Kit produces high-integrity RNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
    • Excellent yield, purity and reproducibility
    • Flexible KingFisher system
    • Walk-away solution for any throughput
    • Compatible with most common downstream analyses and applications
  • Description & Specifications
Related Products
Product Content Feedback