Learn More
Thermo Scientific™ KingFisher™ Pure RNA Tissue Kit
Achieve rapid and reproducible purification of high-quality RNA from cell and tissue samples.
Manufacturer:  Thermo Scientific™ 98040196
 View more versions of this product
Catalog No. 22-387-065
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ KingFisher™ Pure RNA Tissue Kit is designed for rapid automated purification of total RNA from cell and tissue samples using KingFisher instruments. Total RNA can be purified from up to 2 x 106 cells or 20mg of tissue. The excellent performance of KingFisher Pure RNA Tissue Kit produces high-integrity RNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
  • Excellent yield, purity and reproducibility
  • Flexible KingFisher system
  • Walk-away solution for any throughput
  • Compatible with most common downstream analyses and applications
Description & Specifications
Specifications
Tissues or cultured cells
96 samples
KingFisher Pure RNA Tissue Kit; 96 samples
50 min.
Up to 20mg of tissue or 2 x 106 cells
PCR, real-time PCR, enzymatic reactions
100µL