Learn More
Thermo Scientific KingFisher Pure RNA Tissue Kit
Achieve rapid and reproducible purification of high-quality RNA from cell and tissue samples.
Manufacturer: Thermo Scientific 98040496
Description
Thermo Scientific KingFisher Pure RNA Tissue Kit is designed for rapid automated purification of total RNA from cell and tissue samples using KingFisher instruments. Total RNA can be purified from up to 2 x 106 cells or 20mg of tissue. The excellent performance of KingFisher Pure RNA Tissue Kit produces high-integrity RNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific BindIt protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
- Excellent yield, purity and reproducibility
- Flexible KingFisher system
- Walk-away solution for any throughput
- Compatible with most common downstream analyses and applications
Specifications
Tissues or cultured cells | |
384 samples | |
KingFisher Pure RNA Tissue Kit; 384 samples |
Up to 20mg of tissue or 2 x 106 cells | |
PCR, real-time PCR, enzymatic reactions | |
100µL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.