Learn More
Thermo Scientific™ KingFisher™ Pure RNA Tissue Kit
Achieve rapid and reproducible purification of high-quality RNA from cell and tissue samples.
Specifications
Sample Input Up to 20mg of tissue or 2 x 106 cells
Sample Type Tissues or cultured cells
Suitable Downstream Analysis PCR, real-time PCR, enzymatic reactions
Elution Volume (Metric) 100µL
Run Time 50 min.
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
22-387-066
View Documents
Thermo Scientific™
98040496
KingFisher Pure RNA Tissue Kit; 384 samples This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Thermo Scientific™ KingFisher™ Pure RNA Tissue Kit is designed for rapid automated purification of total RNA from cell and tissue samples using KingFisher instruments. Total RNA can be purified from up to 2 x 106 cells or 20mg of tissue. The excellent performance of KingFisher Pure RNA Tissue Kit produces high-integrity RNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
  • Excellent yield, purity and reproducibility
  • Flexible KingFisher system
  • Walk-away solution for any throughput
  • Compatible with most common downstream analyses and applications
Specifications
Specifications
Up to 20mg of tissue or 2 x 106 cells
PCR, real-time PCR, enzymatic reactions
50 min.
Tissues or cultured cells
100µL