Thermo Scientific™ KingFisher™ Pure Viral NA Kit Available on GSA/VA Contract for Federal Government customers only.
Rapidly purify high-quality, reproducible viral RNA and DNA from cell-free body fluids with this unique nucleic acid purification workflow.
Manufacturer: Thermo Scientific™ 98070196
 View more versions of this product
Catalog No. 22-387-071
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Sample Type Cell-free body fluids, such as serum and plasma
    Sample Input 200µL
    Kit Size 96 samples
    Suitable Downstream Analysis PCR, real-time PCR, enzymatic reactions
    Description KingFisher Pure Viral NA Kit; 96 samples
    Elution Volume (Metric) 100µL
    Run Time 40 min.
    Rapidly purify high-quality, reproducible viral RNA and DNA from cell-free body fluids with the Thermo Scientific™ KingFisher™ Pure Viral NA Kit, together with a KingFisher magnetic particle processor.
    Features:
    • Produces viral nucleic acids that are free of inhibitors and ready for use in many downstream applications
    • Facilitate safe sample handling as well as minimal user input, maximizing walkaway time
    • Provides excellent performance and consistency of results
    • Offers superior sensitivity for the purification of viral nucleic acid from cell free body fluids
    • Offers flexible throughput from one sample to up to 96 samples per run
    • Minimizes hands on time, increasing efficiency
    • Compatible with most common downstream analysis and applications
    Recommended for:
    Genomics, target identification, veterinary assays, biomarker discovery, quality control
  • Description & Specifications
Related Products
Product Content Feedback