Learn More
Thermo Scientific™ KingFisher™ Pure Viral NA Kit
Rapidly purify high-quality, reproducible viral RNA and DNA from cell-free body fluids with this unique nucleic acid purification workflow.
Manufacturer:  Thermo Scientific™ 98070496
 View more versions of this product
Catalog No. 22-387-072
  Check Availability
Add to cart
Specifications
Description
Rapidly purify high-quality, reproducible viral RNA and DNA from cell-free body fluids with the Thermo Scientific™ KingFisher™ Pure Viral NA Kit, together with a KingFisher magnetic particle processor.
Features:
  • Produces viral nucleic acids that are free of inhibitors and ready for use in many downstream applications
  • Facilitate safe sample handling as well as minimal user input, maximizing walkaway time
  • Provides excellent performance and consistency of results
  • Offers superior sensitivity for the purification of viral nucleic acid from cell free body fluids
  • Offers flexible throughput from one sample to up to 96 samples per run
  • Minimizes hands on time, increasing efficiency
  • Compatible with most common downstream analysis and applications
Recommended for:
Genomics, target identification, veterinary assays, biomarker discovery, quality control
Description & Specifications
Specifications
Cell-free body fluids, such as serum and plasma
384 samples
KingFisher Pure Viral NA Kit; 384 samples
40 min.
200µL
PCR, real-time PCR, enzymatic reactions
100µL