Thermo Scientific™ KingFisher™ Flex Purification System Available on GSA/VA Contract for Federal Government customers only.

Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Flex Purification Systems.

Manufacturer: Thermo Scientific™ 5400630

 View more versions of this product

Catalog No. 22-387-005

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
    Plates per Deck 24 or 96
    Volume (Metric) Processing 50µL to 1000µL
    Plate Type 24 or 96-well plates:;KingFisher 24 Deep Well plate (200 to 5000µL);Microtiter 96 Deep Well plate (50 to 1000µL);KingFisher 96 plate (20 to 200µL);PCR plate (20 to 100µL)
    Particle Size (Metric) ca. >1µm
    Collection Efficiency of Particles > 95%
    Heating Temperature From +5°C above ambient temperature to +115°C
    Weight (English) 62 lb.
    Weight (Metric) 28kg
    Capacity 96 samples
    Depth (English) 23.6 in.
    Depth (Metric) 60cm
    Description KingFisher 96 Plate, KingFisher 96 DW plate
    Display START / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD
    For Microplate Type Microtiter 96 deep-well plate (50 to 1000µL), KingFisher 96 KF plate (50-150 µl)
    Height (English) 15 in.
    Height (Metric) 38cm
    Item Description KingFisher Flex, with 96 Deep-well Head
    Magnetic Rods 96 in one frame; Interchangeable magnet heads
    Width (English) 26.8 in.
    Width (Metric) 68cm

    Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Flex Purification Systems, which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.

    Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.

    • Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
    • High throughput system processes up to 96 samples in less than 20 minutes.
    • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
    • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
    • Ready-made protocols for different type of applications are available.

    Recommended for:

    • Genomics and proteomics
    • Target identification
    • Veterinary assays
    • Biomarker discovery
    • Quality control
    • Forensics

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit