Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kininogen-1 Armenian Hamster anti-Human, Clone: 2B7, MilliporeSigma™

Armenian Hamster Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen Kininogen-1
Clone 2B7
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN2630100
View Documents
MilliporeSigma
MABN2630100UG
100 μg
Each for $387.00
Add to cart
 
MABN263025
View Documents
MilliporeSigma
MABN263025UG
25 μg
Each for $153.40
Add to cart
 
Description

Description

Kininogen-1 Monoclonal antibody specifically detects Kininogen-1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation.
Specifications

Specifications

Kininogen-1
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Purified
RUO
Purified armenian Hamster monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Monoclonal
Protein G purified
2B7
Unconjugated
Armenian Hamster
Human
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Kininogen-1.
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit